Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylene Blue, 1% w/v, Reagents
Click to view available options
Quantity:
120 mL
500 mL
Specifications
Specifications
Chemical Name or Material | Methylene Blue, 1% w/v |
CAS | 7732-18-5, 7220-79-3 |
Boiling Point | 100°C |
Synonym | 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride Trihydrate, Basic Blue 9 Trihydrate, Color Index 52015 |
Percent Purity | 1% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.005 |
Color | Blue |
Quantity | 120 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction