Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
25g
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7220-79-3
C16H24ClN3O3S
373.896
XQAXGZLFSSPBMK-UHFFFAOYSA-M
C.I. 52015
104827

High quality chemicals for laboratory and specialized industrial use

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Synonym C.I. 52015
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.O.O.O.[Cl-]
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
Molecular Weight (g/mol) 373.896
PubChem CID 104827
Formula Weight 373.91
Packaging Wide Mouth Amber Glass Bottle
Quantity 100g
Chemical Name or Material Methylene Blue
Grade BAKER ANALYZED™ Reagent
Show More Show Less