Methylene Blue Solution II, 0.05mL (1 drop) = 0.1mg/L Sulfide, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 491016

Packaging Poly natural
Quantity 500mL

 View more versions of this product

Catalog No. 4910-16

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7732-18-5,7220-79-3
    CAS Min % 98.0,0.01
    CAS Max % 100.0,0.01
    Physical Form Liquid
    Chemical Name or Material Methylene Blue Solution II

    Specifications

    Physical Form Liquid
    Color Clear blue
    Quantity 500mL
    Equivalence 0.05 mL (1 drop) = 0.1 mg/L Sulfide
    CAS 7732-18-5,7220-79-3
    CAS Min % 98.0,0.01
    Packaging Poly natural
    CAS Max % 100.0,0.01
    Chemical Name or Material Methylene Blue Solution II

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit