Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl

High quality chemicals for laboratory and specialized industrial use

Formula Weight 84.93
Physical Form Liquid
Grade Meets ACS reagent chemical requirements
Density 1.318g/mL at 25°C (typical)
Density at 25 degC 1.318g/mL
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.32kg
Boiling Point 39.5°C (typical)
Color Colorless
Quantity 4L
Chemical Name or Material Methylene Chloride
Show More Show Less