Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl

High quality chemicals for laboratory and specialized industrial use

Formula Weight 84.93
Physical Form Liquid
Grade Meets ACS reagent chemical requirements
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.32kg
Quantity 500mL
Chemical Name or Material Methylene Chloride