Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, GCSolv (TM), 99.9%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
75-09-2
75-09-2
CAS Max % 1
Percent Purity 0.999
Grade High Purity Solvent
Chloride 10 ppm
Packaging Amber Glass Bottle
Residue after Evaporation 1 ppm
Titratable Acid 0.0003 meq/g
Water 0.0001
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.