Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle

Chemical Identifiers

75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
CHEBI:15767

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry; For laboratory, research, or manufacturing use
CAS 75-09-2
Molecular Formula CH2Cl2
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES C(Cl)Cl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.927
PubChem CID 6344
ChEBI CHEBI:15767
Formula Weight 84.93
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Specific Gravity 1L = 1.32kg
Quantity 4L
Chemical Name or Material Methylene Chloride
Grade HPLC Grade
Show More Show Less