Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry; For laboratory, research, or manufacturing use
Formula Weight 84.93
Grade HPLC
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.32kg
Quantity 1L
Chemical Name or Material Methylene Chloride