Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, Low water, BakerDRY™, J.T.Baker™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum

Chemical Identifiers

75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
CHEBI:15767

Specifically manufactured for use in organometallic and oligonucleotide synthesis

CAS 75-09-2
Molecular Formula CH2Cl2
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES C(Cl)Cl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.927
PubChem CID 6344
ChEBI CHEBI:15767
Formula Weight 84.93
Packaging Glass Bottle
Specific Gravity 1L = 1.32kg
Quantity 100mL
Chemical Name or Material Methylene Chloride, Low water
Grade BakerDRY™
Show More Show Less