Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, Low water, BakerDRY™, J.T.Baker™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl

Specifically manufactured for use in organometallic and oligonucleotide synthesis

Formula Weight 84.93
Grade BakerDRY™
Packaging Narrow Mouth Amber Glass Bottle, w/ Septum
Specific Gravity 1L = 1.32kg
Quantity 1L
Chemical Name or Material Methylene Chloride, Low water