Alfa Aesar™ Methylene dithiocyanate, 98%

Manufacturer: Alfa Aesar™ A1074018

Quantity 50g

 View more versions of this product

Catalog No. AAA1074018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6317-18-6
    Molecular Formula CH2(SCN)2
    Formula Weight 130.19
    MDL Number MFCD00001833
    UN Number UN2928
    Synonym Dithiocyanatomethane
    Chemical Name or Material Methylene dithiocyanate

    Specifications

    Formula Weight 130.19
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula CH2(SCN)2
    CAS 6317-18-6
    UN Number UN2928
    MDL Number MFCD00001833
    Synonym Dithiocyanatomethane
    Health Hazard 1 Danger
    Melting Point 103° to 107°C
    Quantity 50g
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H301-H330-H314-H317
    Toxic if swallowed.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    May cause an allergic skin reaction.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    RTECS Number XL1560000
    TSCA TSCA
    Chemical Name or Material Methylene dithiocyanate
    Assay 98%

    Description

    Applications
    Methylene dithiocyanate is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field. It is also used to control slime in pulp and paper manufacturing, preservation of paints, adhesives, synthetic polymer lattices and latex emulsions, anti-microbial treatment of water thinned products, thickeners and slurries, preservation of wood and wood products, preservation of Leather and Leather products and in oil well brines and drilling mud.

    Solubility
    Decomposes in water. Slightly soluble in organic solvents.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H330-H314-H317
    Toxic if swallowed.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit