Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl picolinate, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
2459-07-6
C7H7NO2
137.14
NMMIHXMBOZYNET-UHFFFAOYSA-N
methyl picolinate,2-picolinic acid methyl ester,2-pyridinecarboxylic acid, methyl ester,methyl 2-pyridinecarboxylate,picolinic acid, methyl ester,2-carbomethoxypyridine,pyridine-2-carboxylic acid methyl ester,2-pyridinecarboxylic acid methyl ester,picolinic acid methyl ester,methylpicolinate
75569
methyl pyridine-2-carboxylate
COC(=O)C1=CC=CC=N1
2459-07-6
C7H7NO2
137.14
NMMIHXMBOZYNET-UHFFFAOYSA-N
methyl picolinate,2-picolinic acid methyl ester,2-pyridinecarboxylic acid, methyl ester,methyl 2-pyridinecarboxylate,picolinic acid, methyl ester,2-carbomethoxypyridine,pyridine-2-carboxylic acid methyl ester,2-pyridinecarboxylic acid methyl ester,picolinic acid methyl ester,methylpicolinate
75569
methyl pyridine-2-carboxylate
COC(=O)C1=CC=CC=N1
Melting Point 14°C to 16°C
Boiling Point 95°C (1.0 hPa)
Flash Point 103°C
Specific Gravity 1.137
Density 1.1370g/mL
Quantity 100g
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Formula Weight 137.14
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Methyl picolinate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 219-545-2