Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl pyruvate, 94%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
600-22-6
C4H6O3
102.09
MFCD00008754
CWKLZLBVOJRSOM-UHFFFAOYSA-N
methyl pyruvate,pyruvic acid, methyl ester,pyruvic acid methyl ester,methyl 2-oxopropionate,propanoic acid, 2-oxo-, methyl ester,methyl-pyruvate,methylglyoxylic acid methyl ester,unii-3kjm65g5xl,3kjm65g5xl,2-oxo-propionic acid methyl ester
11748
CHEBI:51850
methyl 2-oxopropanoate
CC(=O)C(=O)OC
600-22-6
C4H6O3
102.09
MFCD00008754
CWKLZLBVOJRSOM-UHFFFAOYSA-N
methyl pyruvate,pyruvic acid, methyl ester,pyruvic acid methyl ester,methyl 2-oxopropionate,propanoic acid, 2-oxo-, methyl ester,methyl-pyruvate,methylglyoxylic acid methyl ester,unii-3kjm65g5xl,3kjm65g5xl,2-oxo-propionic acid methyl ester
11748
CHEBI:51850
methyl 2-oxopropanoate
CC(=O)C(=O)OC
Specific Gravity 1.13
Boiling Point 134.0°C to 137.0°C
Refractive Index 1.4065
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
Density 1.1300g/mL
Flash Point 39°C
Quantity 500g
Assay Percent Range 94%
Linear Formula CH3COCO2CH3
Beilstein 03, 616
Merck Index 15, 8135
Solubility Information Solubility in water: soluble.
Formula Weight 102.09
Physical Form Liquid
Percent Purity 94%
Chemical Name or Material Methyl pyruvate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning