Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Thioglycolate, 95%, Thermo Scientific Chemicals

Catalog Number AC148881000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2365-48-2
C3H6O2S
106.14
MFCD00004873
MKIJJIMOAABWGF-UHFFFAOYSA-N
methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el
16907
methyl 2-sulfanylacetate
COC(=O)CS
2365-48-2
C3H6O2S
106.14
MFCD00004873
MKIJJIMOAABWGF-UHFFFAOYSA-N
methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el
16907
methyl 2-sulfanylacetate
COC(=O)CS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -24°C
Color Colorless
Density 1.1870g/mL
Boiling Point 156°C to 159°C (10.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Iodimetry)
Packaging Glass bottle
Refractive Index 1.4640 to 1.467
Linear Formula HSCH2CO2CH3
Quantity 100 g
Specific Gravity 1.187
Solubility Information Solubility in water: 40g/l water (20°C)
Formula Weight 106.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl Thioglycolate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-121-7

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit