Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Thioglycolate, 95%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
2365-48-2
C3H6O2S
106.14
MFCD00004873
MKIJJIMOAABWGF-UHFFFAOYSA-N
methyl thioglycolate,methyl mercaptoacetate,methyl 2-mercaptoacetate,methylthioglycolate,thioglycolic acid methyl ester,acetic acid, mercapto-, methyl ester,methanethiol, methoxy,usaf ek-7119,thioglykolsaeure-methylester,unii-o1608la9el
16907
methyl 2-sulfanylacetate
COC(=O)CS
2365-48-2
C3H6O2S
106.14
MFCD00004873
MKIJJIMOAABWGF-UHFFFAOYSA-N
methyl thioglycolate,methyl mercaptoacetate,methyl 2-mercaptoacetate,methylthioglycolate,thioglycolic acid methyl ester,acetic acid, mercapto-, methyl ester,methanethiol, methoxy,usaf ek-7119,thioglykolsaeure-methylester,unii-o1608la9el
16907
methyl 2-sulfanylacetate
COC(=O)CS
Melting Point -24°C
Boiling Point 156°C to 159°C (10.0mmHg)
Flash Point 56°C
Specific Gravity 1.187
Density 1.1870g/mL
Refractive Index 1.4640 to 1.467
Color Colorless
Quantity 500g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 95%
Linear Formula HSCH2CO2CH3
Solubility Information Solubility in water: 40g/l water (20°C)
Formula Weight 106.14
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material Methyl Thioglycolate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger