Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyletrimethylacetate, 99%, ACROS Organics™

Click to view available options
Quantity:
100mL
250mL
Packaging:
Glass bottle
598-98-1
C6H12O2
116.16
MFCD00008843
CNMFHDIDIMZHKY-UHFFFAOYSA-N
methyl pivalate,methyl trimethylacetate,propanoic acid, 2,2-dimethyl-, methyl ester,pivalic acid, methyl ester,unii-bfx9w386ox,bfx9w386ox,pivalic acid methyl ester,methyl pivaloate,methyltrimethylacetate,tert-c4h9cooch3
69027
methyl 2,2-dimethylpropanoate
CC(C)(C)C(=O)OC
598-98-1
C6H12O2
116.16
MFCD00008843
CNMFHDIDIMZHKY-UHFFFAOYSA-N
methyl pivalate,methyl trimethylacetate,propanoic acid, 2,2-dimethyl-, methyl ester,pivalic acid, methyl ester,unii-bfx9w386ox,bfx9w386ox,pivalic acid methyl ester,methyl pivaloate,methyltrimethylacetate,tert-c4h9cooch3
69027
methyl 2,2-dimethylpropanoate
CC(C)(C)C(=O)OC
Melting Point -70°C
Boiling Point 101°C
Flash Point <1°C
Viscosity 30 mPa.s (25°C)
Specific Gravity 0.87
Refractive Index 1.3890 to 1.391
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless
Quantity 250mL
Density 0.8700g/mL
Assay Percent Range 99%
Linear Formula (CH3)3CCO2CH3
Beilstein 02, 320
Formula Weight 116.16
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Methyl trimethylacetate
Show More Show Less

GHS P Statement Highly flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

GHS Signal Word: Danger