Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Methylferrocenemethanol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1277-49-2
C12 H14 Fe O
MFCD00013752
1-(Ferrocenyl)ethanol
1277-49-2
C12 H14 Fe O
MFCD00013752
1-(Ferrocenyl)ethanol
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Orange-Yellow to Yellow
Melting Point 75°C to 80°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 230.09
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material α-Methylferrocenemethanol, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.