Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylglyoxal Monoclonal Antibody (9F11), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527568
Description
Methylglyoxal Monoclonal antibody specifically detects Methylglyoxal in Chemical samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
Methylglyoxal | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
-20°C | |
Liquid | |
Chemical |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
9F11 | |
Unconjugated | |
2 oxo propanal; 2-Oxopropanal; Aldoketomutase; epididymis secretory protein Li 74; GLO1; GLOD1; Glx I; GLYI; glyoxalase domain containing 1; glyoxalase I; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; Methylglyoxal; Methylglyoxal (MG); Methylglyoxalase; MG; MG-modified protein; Pyruvaldehyde; S-D-lactoylglutathione methylglyoxal lyase | |
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH) | |
100 μg | |
Primary | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction