Learn More
Methylglyoxal Mouse anti-Human, Non-species specific, DyLight 755, Clone: 90000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259368IR
Catalog No. NBP259368IR
Add to cart
Description
Description
Methylglyoxal Monoclonal antibody specifically detects Methylglyoxal in Human, Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Methylglyoxal
90000000
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Purified
GLO1
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Non-species specific
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
2-oxopropanal, Pyruvaldehyde
Mouse
IgG2a
100 ul
Primary
Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins.