Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylglyoxal Mouse anti-Human, Nonspecies specific, Allophycocyanin/Cy7, Clone: 90000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259368APCCY7

Catalog No. N259368APC7


Add to cart

Description

Description

Methylglyoxal Monoclonal antibody specifically detects Methylglyoxal in Human, Nonspecies specific samples. It is validated for Flow Cytometry.
Specifications

Specifications

Methylglyoxal
90000000
Flow Cytometry
Conjugated, Purified
Mouse
IgG2a
0.1 mL
Primary
Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins.
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
2-oxopropanal, Pyruvaldehyde
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Nonspecies specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit