Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Methylhydrocinnamic acid, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1009-67-2
C10H12O2
164.2
MFCD00192301
MCIIDRLDHRQKPH-UHFFFAOYSA-N
2-benzylpropionic acid,alpha-methylhydrocinnamic acid,2-methyl-3-phenylpropionic acid,2-benzylpropanoic acid,2-methyl-3-phenyl-propionic acid,acmc-20ao3h,alpha-methylhydrocinnamicacid,alpha-methyl-hydrocinnamic acid,2-methyl-3-phenyl-propanoic acid,#
99862
2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)C(=O)O
1009-67-2
C10H12O2
164.2
MFCD00192301
MCIIDRLDHRQKPH-UHFFFAOYSA-N
2-benzylpropionic acid,alpha-methylhydrocinnamic acid,2-methyl-3-phenylpropionic acid,2-benzylpropanoic acid,2-methyl-3-phenyl-propionic acid,acmc-20ao3h,alpha-methylhydrocinnamicacid,alpha-methyl-hydrocinnamic acid,2-methyl-3-phenyl-propanoic acid,#
99862
2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)C(=O)O
Boiling Point 167°C to 168°C (23.0mmHg)
Flash Point 113°C
Packaging Glass bottle
Quantity 1g
Melting Point 37°C to 41°C
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula C6H5CH2CH(CH3)CO2H
Formula Weight 164.2
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material α-Methylhydrocinnamic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning