Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Methylhydrocinnamic acid, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1009-67-2
C10H12O2
164.2
MFCD00192301
MCIIDRLDHRQKPH-UHFFFAOYSA-N
2-benzylpropionic acid,alpha-methylhydrocinnamic acid,2-methyl-3-phenylpropionic acid,2-benzylpropanoic acid,2-methyl-3-phenyl-propionic acid,acmc-20ao3h,alpha-methylhydrocinnamicacid,alpha-methyl-hydrocinnamic acid,2-methyl-3-phenyl-propanoic acid,#
99862
2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)C(=O)O
1009-67-2
C10H12O2
164.2
MFCD00192301
MCIIDRLDHRQKPH-UHFFFAOYSA-N
2-benzylpropionic acid,alpha-methylhydrocinnamic acid,2-methyl-3-phenylpropionic acid,2-benzylpropanoic acid,2-methyl-3-phenyl-propionic acid,acmc-20ao3h,alpha-methylhydrocinnamicacid,alpha-methyl-hydrocinnamic acid,2-methyl-3-phenyl-propanoic acid,#
99862
2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)C(=O)O
Boiling Point 167°C to 168°C (23.0mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Melting Point 37°C to 41°C
Packaging Glass bottle
Quantity 5g
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5CH2CH(CH3)CO2H
Formula Weight 164.2
Physical Form Crystalline Powder or Crystals
Percent Purity ≥97.5%
Chemical Name or Material α-Methylhydrocinnamic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

;GHS P StatementIF ON SKIN: Wash with plenty of soap and water.If eye irritation persists: Get medical advice/attention.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/