Alfa Aesar™ Methyllithium, 1-2M in ether

Used in Grignard-type reactions

Manufacturer: Alfa Aesar™ 89693AE

Quantity 100mL

 View more versions of this product

Catalog No. AA89693AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 917-54-4
    Formula Weight 21.97
    MDL Number MFCD00008253
    UN Number UN3394
    Chemical Name or Material Methyllithium

    Specifications

    Formula Weight 21.97
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    Health Hazard 2 GHS H Statement
    H224-H250-H260-H252-H314-H318-H302-H336
    Extremely flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.
    MDL Number MFCD00008253
    Odor Ether-like
    DOT Information Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
    CAS 917-54-4
    UN Number UN3394
    Density 0.732g/mL
    Health Hazard 1 Danger
    Color Colorless
    Sensitivity Air & Moisture Sensitive
    Concentration 1 to 2M in ether
    TSCA TSCA
    Physical Form Liquid
    Quantity 100mL
    Chemical Name or Material Methyllithium

    GHS H Statement
    H224-H250-H260-H252-H314-H318-H302-H336
    Extremely flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit