Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyllithium, 2.2M (6wt%) in diethyl ether with LiBr, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL,100mL
800mL,800mL
Packaging:
AcroSeal™ Glass Bottle,AcroSeal™ Glass Bottle
917-54-4,60-29-7
CH3Li
21.98
MFCD00044645
IHLVCKWPAMTVTG-UHFFFAOYSA-N
methyllithium,lithium, methyl,methyl lithium,lithium methanide,meli,lithium methyl,lithium carbanide,lithium methide,methllithium,methyllithum
2724049
lithium;carbanide
[Li+].[CH3-]
917-54-4,60-29-7
CH3Li
21.98
MFCD00044645
IHLVCKWPAMTVTG-UHFFFAOYSA-N
methyllithium,lithium, methyl,methyl lithium,lithium methanide,meli,lithium methyl,lithium carbanide,lithium methide,methllithium,methyllithum
2724049
lithium;carbanide
[Li+].[CH3-]
Packaging AcroSeal™ Glass Bottle,AcroSeal™ Glass Bottle
Flash Point −28°C,−28°C
Specific Gravity 0.81,0.81
Color Colorless to Yellow,Colorless through Yellow
Quantity 800mL,800mL
Density 0.8100g/mL,0.8100g/mL
Linear Formula CH3Li
Solubility Information Solubility in water: vigorous reaction
Formula Weight 21.98
Physical Form Solution
Percent Purity 97%
Storage Information Product may develop some turbidity or precipitate
Chemical Name or Material Methyllithium
Show More Show Less

GHS H Statement
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 213-026-4

Recommended Storage : Product may develop some turbidity or precipitate