Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyllycaconitine citrate, Tocris Bioscience™
Click to view available options
Quantity:
5 mg
25 mg
Description
Potent antagonist for α7-containing neuronal nicotinic receptors (Ki = 1.4 nM). Interacts with α4β2 and α6β2 receptors at concentrations > 40 nM. Attenuates METH-induced neurotoxicity in mouse striatum in vivo.
Specifications
Quantity | 25 mg |
Formula Weight | 874.93 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | Methyllycaconitine citrate |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction