Alfa Aesar™ Methylmagnesium bromide, 3M in ether

Manufacturer: Alfa Aesar™ 87324JU

Quantity 11 mole

 View more versions of this product

Catalog No. AA87324JU

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 75-16-1
    Linear Formula CH3MgBr
    Formula Weight 119.26
    MDL Number MFCD00000041
    UN Number UN1928
    Chemical Name or Material Methylmagnesium bromide

    Specifications

    Formula Weight 119.26
    Health Hazard 2 GHS H Statement
    H224-H261-H314-H318-H302-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    Density 1.035g/mL
    Physical Form Liquid
    MDL Number MFCD00000041
    CAS 75-16-1
    Color Colorless to pale brown
    Flash Point −40°C (−40°F)
    Health Hazard 1 Danger
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METHYL MAGNESIUM BROMIDE, IN ETHYL ETHER
    TSCA TSCA
    Linear Formula CH3MgBr
    UN Number UN1928
    Quantity 11 mole
    Odor Ether-like
    Concentration 3M in ether
    Chemical Name or Material Methylmagnesium bromide

    GHS H Statement
    H224-H261-H314-H318-H302-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit