Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 41252JS

Quantity 0.25 mole

 View more versions of this product

Catalog No. AA41252JS

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
Methylmagnesium bromide, 3M in ether has the wide range of applications in organic synthesis. Alkylation of aldehydes and ketones is the most common applications of Grignard reagent. It is a strong nucleophile that can form new carbon-carbon bonds. Also like organolithium compounds, it is useful to form carbon-heteroatom bonds.

Solubility
Soluble in ether, toluene and tetrahydrfuran. Insoluble in hydrocarbons.

Notes
It is highly flammable and reacts with moisture or water to form methane gas, so it should be stored in tightly closed containers.
Description & Specifications

Chemical Identifiers

75-16-1
119.26
UN1928
CH3MgBr
MFCD00000041
Methylmagnesium bromide

Specifications

75-16-1
ChemSeal™ bottles
CH3MgBr
MFCD00000041
Pale brown
−40° (−40°F)
3M in ether
Methylmagnesium bromide
1.035g/mL
Liquid
119.26
UN1928
0.25 mole
Ether-like
Air & Moisture Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H224-H261-H314-H318-H302-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METHYL MAGNESIUM BROMIDE, IN ETHYL ETHER

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit