Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methylmagnesium chloride, 3M in THF, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 42101JU

Quantity 11 mole

 View more versions of this product

Catalog No. AA42101JU


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

676-58-4
74.81
UN3399
Methylmagnesium chloride
CH3MgCl
MFCD00000468
3587199

Specifications

676-58-4
ChemSeal™ bottles
CH3MgCl
MFCD00000468
3587199
3M in THF
−17° (1°F)
Air & Moisture Sensitive
1.013g/mL
Liquid
74.81
UN3399
Pale brown to dark green
11 mole
Ether-like
Methylmagnesium chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit