Alfa Aesar™ Methylmagnesium chloride, 3M in THF, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 42101JS

Quantity 0.25 mole

 View more versions of this product

Catalog No. AA42101JS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 676-58-4
    Linear Formula CH3MgCl
    Formula Weight 74.81
    MDL Number MFCD00000468
    UN Number UN3399
    Beilstein 3587199
    Chemical Name or Material Methylmagnesium chloride

    Specifications

    Formula Weight 74.81
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    UN Number UN3399
    Color Pale brown to dark green
    Flash Point −17° (1°F)
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    Density 1.013g/mL
    Physical Form Liquid
    CAS 676-58-4
    Beilstein 3587199
    Health Hazard 1 Danger
    Sensitivity Air & Moisture Sensitive
    TSCA TSCA
    Linear Formula CH3MgCl
    MDL Number MFCD00000468
    Quantity 0.25 mole
    Storage Note 1
    • Packaged under argon
    Packaging ChemSeal™ bottles
    Odor Ether-like
    Health Hazard 2 GHS H Statement
    H225-H261-H314-H318
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Concentration 3M in THF
    Chemical Name or Material Methylmagnesium chloride

    GHS H Statement
    H225-H261-H314-H318
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit