Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylmalonic acid, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
516-05-2
C4H6O4
118.09
MFCD00002656
ZIYVHBGGAOATLY-UHFFFAOYSA-N
methylmalonic acid,2-methylmalonic acid,isosuccinic acid,methylmalonate,propanedioic acid, methyl,1,1-ethanedicarboxylic acid,malonic acid, methyl,methyl malonic acid,methylpropanedioic acid,unii-8ll8s712j7
487
CHEBI:30860
2-methylpropanedioic acid
CC(C(=O)O)C(=O)O
516-05-2
C4H6O4
118.09
MFCD00002656
ZIYVHBGGAOATLY-UHFFFAOYSA-N
methylmalonic acid,2-methylmalonic acid,isosuccinic acid,methylmalonate,propanedioic acid, methyl,1,1-ethanedicarboxylic acid,malonic acid, methyl,methyl malonic acid,methylpropanedioic acid,unii-8ll8s712j7
487
CHEBI:30860
2-methylpropanedioic acid
CC(C(=O)O)C(=O)O
Packaging Glass bottle
Density 1.4500g/mL
Melting Point 129.0°C to 136.0°C
Specific Gravity 1.45
Quantity 5g
Infrared Spectrum Authentic
Color White
Water 0.5% max. (K.F.)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula HO2CCH(CH3)CO2H
Beilstein 02, IV, 1932
Solubility Information Solubility in water: very soluble. Other solubilities: very soluble in ethanol,soluble in ether and ethyl acetate,slightly soluble in benzene and trifluoroacetic,acid
Formula Weight 118.09
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material Methylmalonic acid
Show More Show Less