Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylmalonyl-L-Carnitine Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™

Manufacturer:  MilliporeSigma Supelco 9518710MG

Catalog No. 11-102-2820


Add to cart

Description

Description

Specifications

Specifications

251976-17-7 (Free Acid)
C11H19NO6 · xLi+
261.27 (Free Acid Basis)
Analytical Standard
10 mg
C11H19NO6 · xLi+
(2 R)-3-Carboxy-2-(2-carboxy-1-oxopropoxy)-N,N,N-trimethyl-1-propanaminium inner salt lithium salt; O-Methylmalonyl-L-carnitine lithium salt
≥90% (HPLC)
Not Applicable
[α]/D -33°C ± 2°C = 0.1 in. H2O
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

Recommended Storage : 2°C to 8°C

SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit