Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylmercury(II) chloride, standard solution in H2O, Alfa Aesar™
Click to view available options
Quantity:
5mL
25mL
Specifications
Beilstein | 3600218 |
Appearance | Clear |
Solubility Information | Miscible with lipid. |
Formula Weight | 251.08 |
Physical Form | Liquid |
Color | Undesignated |
Quantity | 5mL |
Chemical Name or Material | Methylmercury(II) chloride |
Safety and Handling
P260-P314-P501c
H373
DOTInformation : DOT Class: 6.1; Packing Group: II
EINECSNumber : 204-064-2
TSCA : No
Recommended Storage : Ambient temperatures