Methylmercury(II) chloride, standard solution in H2O, Alfa Aesar™

Manufacturer: Alfa Aesar 33553AC

Quantity 25mL

 View more versions of this product

Catalog No. AA33553AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 115-09-3
    Physical Form Liquid
    Molecular Formula CH3HgCl
    Formula Weight 251.08
    MDL Number MFCD00013592
    Synonym Chloromethylmercury
    Chemical Name or Material Methylmercury(II) chloride

    Specifications

    Formula Weight 251.08
    Quantity 25mL
    Color Clear
    DOT Information DOT Class: 6.1; Packing Group: II
    EINECS Number 204-064-2
    Physical Form Liquid
    Molecular Formula CH3HgCl
    Appearance Clear
    MDL Number MFCD00013592
    CAS 115-09-3
    Synonym Chloromethylmercury
    Health Hazard 1 H300-H310-H330-H373-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Methylmercury(II) chloride

    H300-H310-H330-H373-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit