Alfa Aesar™ Methylmercury(II) hydroxide, 1M in H2O, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 41253AD

Quantity 50mL

 View more versions of this product

Catalog No. AA41253AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1184-57-2
    Linear Formula CH3HgOH
    Formula Weight 232.63
    MDL Number MFCD00013924
    UN Number UN2024
    Chemical Name or Material Methylmercury(II) hydroxide

    Specifications

    Formula Weight 232.63
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Packaging ChemSeal™ bottles
    Color Colorless
    Health Hazard 1 Danger
    Quantity 50mL
    Storage Note 1
    • Packaged under argon
    Physical Form Liquid
    Linear Formula CH3HgOH
    MDL Number MFCD00013924
    UN Number UN2024
    Concentration 1M in H2O
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, LIQUID, N.O.S.
    CAS 1184-57-2
    Health Hazard 2 GHS H Statement
    H300-H311-H331-H373
    Fatal if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    RTECS Number OW4900000
    Chemical Name or Material Methylmercury(II) hydroxide

    GHS H Statement
    H300-H311-H331-H373
    Fatal if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P261-P301+P310-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit