Learn More
Medchemexpress LLC Methylophiopogonone B | 74805-89-3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN24421MG
Encompass_Preferred
Methylophiopogonone B is a homoisoflavonoidal compound isolated from Ophiopogonis Tuber. It demonstrates the ability to scavenge hydroxyl radicals (•OH) and hydrogen peroxide (H2O2) in vitro, making it suitable for various research applications.
  • Homoisoflavonoidal compound
  • Isolated from Ophiopogonis Tuber
  • Capable of scavenging hydroxyl radicals and hydrogen peroxide in vitro
  • Identified for use as laboratory chemical and in substance manufacturing
  • Targets reactive oxygen species
  • Involves pathways in immunology/inflammation, metabolic enzyme/protease, and NF-κB
  • For research use only
  • Stable under recommended storage conditions
  • Appearance: solid, off-white to light yellow
Catalog No. 50-004-21849