Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methylphenylphosphinic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
4271-13-0
C7H9O2P
156.121
MFCD00123313
RMJCJLHZCBFPDN-UHFFFAOYSA-N
methyl phenyl phosphinic acid,methyl phenylphosphinic acid,acmc-20akeb,?-methylphenylphosphinic acid,methylphenylphosphinic acid
345468
methyl(phenyl)phosphinic acid
CP(=O)(C1=CC=CC=C1)O
4271-13-0
C7H9O2P
156.121
MFCD00123313
RMJCJLHZCBFPDN-UHFFFAOYSA-N
methyl phenyl phosphinic acid,methyl phenylphosphinic acid,acmc-20akeb,?-methylphenylphosphinic acid,methylphenylphosphinic acid
345468
methyl(phenyl)phosphinic acid
CP(=O)(C1=CC=CC=C1)O
Quantity 25g
Melting Point 132°C to 134°C
Formula Weight 156.12
Percent Purity 98%
Chemical Name or Material Methylphenylphosphinic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures