Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methylphenylphosphinic chloride, 97%

Click to view available options
Quantity:
1g
5g
25g
5761-97-7
C7H8ClOP
174.564
MFCD00013902
BTMQENOUJJKBGT-UHFFFAOYSA-N
methylphenylphosphinic chloride,chloro methyl phosphoryl benzene,methyl phenyl phosphinoyl chloride,acmc-1arvk,methyl phenylphosphinicchloride,methylphenylphosphinyl chloride,methylphenyl-phosphinic chloride,phenyl methyl phosphinyl chloride,phosphinic chloride,methylphenyl
21983
[chloro(methyl)phosphoryl]benzene
CP(=O)(C1=CC=CC=C1)Cl
5761-97-7
C7H8ClOP
174.564
MFCD00013902
BTMQENOUJJKBGT-UHFFFAOYSA-N
methylphenylphosphinic chloride,chloro methyl phosphoryl benzene,methyl phenyl phosphinoyl chloride,acmc-1arvk,methyl phenylphosphinicchloride,methylphenylphosphinyl chloride,methylphenyl-phosphinic chloride,phenyl methyl phosphinyl chloride,phosphinic chloride,methylphenyl
21983
[chloro(methyl)phosphoryl]benzene
CP(=O)(C1=CC=CC=C1)Cl
Density 1.266
Sensitivity Moisture sensitive
Boiling Point 155°C (11mmHg)
Quantity 1g
Flash Point >110°C (230°F)
UN Number UN3261
Formula Weight 174.57
Percent Purity 97%
Chemical Name or Material Methylphenylphosphinic chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Keep cold