Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylphenylvinylsilane, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
17878-39-6
C9H12Si
148.28
MFCD00053689
MOSXLDGILGBOSZ-UHFFFAOYSA-N
methylphenylvinylsilane,silane, ethenylmethylphenyl,phenylmethylvinylsilane,benzene, ethenylmethylsilyl,methyl phenyl vinylsilane,ethenyl methyl phenylsilyl,phenyl methyl vinyl silane,methyl phenyl vinyl silane,methyl phenyl vinylsilane #,vinylphenylmethylsilane
6329252
ethenyl-methyl-phenylsilicon
C[Si](C=C)C1=CC=CC=C1
17878-39-6
C9H12Si
148.28
MFCD00053689
MOSXLDGILGBOSZ-UHFFFAOYSA-N
methylphenylvinylsilane,silane, ethenylmethylphenyl,phenylmethylvinylsilane,benzene, ethenylmethylsilyl,methyl phenyl vinylsilane,ethenyl methyl phenylsilyl,phenyl methyl vinyl silane,methyl phenyl vinyl silane,methyl phenyl vinylsilane #,vinylphenylmethylsilane
6329252
ethenyl-methyl-phenylsilicon
C[Si](C=C)C1=CC=CC=C1
Reduction reagent
Packaging Glass bottle
Density 0.8900g/mL
Refractive Index 1.5125 to 1.5145
Specific Gravity 0.89
Boiling Point 56°C to 57°C (7.0mmHg)
Infrared Spectrum Authentic
Quantity 5g
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula C6H5Si(H)(CH3)CHCH2
Solubility Information Solubility in water: solubility in water: negligible
Formula Weight 148.28
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Methylphenylvinylsilane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 241-830-5

TSCA : TSCA