Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (Methylthio)acetaldehyde dimethyl acetal, 97%

Click to view available options
Quantity:
5g
25g
40015-15-4
C5H12O2S
136.209
MFCD00009848
DVOAQLUDKIFSNB-UHFFFAOYSA-N
methylthio acetaldehyde dimethyl acetal,1,1-dimethoxy-2-methylthio ethane,ethane, 1,1-dimethoxy-2-methylthio,1,1-dimethoxy-2-methylsulfanyl-ethane,ethane,1,1-dimethoxy-2-methylthio,methylmercapto acetaldehyde dimethyl acetal,1,1-dimethoxy-2-methylsulfanyl ethane,acmc-209j9v,methylthio acetaldehyde dimethylacetal,2-methylthio acetaldehyde dimethyl acetal
638113
1,1-dimethoxy-2-methylsulfanylethane
COC(CSC)OC
40015-15-4
C5H12O2S
136.209
MFCD00009848
DVOAQLUDKIFSNB-UHFFFAOYSA-N
methylthio acetaldehyde dimethyl acetal,1,1-dimethoxy-2-methylthio ethane,ethane, 1,1-dimethoxy-2-methylthio,1,1-dimethoxy-2-methylsulfanyl-ethane,ethane,1,1-dimethoxy-2-methylthio,methylmercapto acetaldehyde dimethyl acetal,1,1-dimethoxy-2-methylsulfanyl ethane,acmc-209j9v,methylthio acetaldehyde dimethylacetal,2-methylthio acetaldehyde dimethyl acetal
638113
1,1-dimethoxy-2-methylsulfanylethane
COC(CSC)OC
Quantity 5g
Flash Point 53°C (127°F)
Boiling Point 170°C to 171°C
UN Number UN1993
Beilstein 773689
Formula Weight 136.21
Percent Purity 97%
Chemical Name or Material (Methylthio)acetaldehyde dimethyl acetal

GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.