Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Methylthio)acetaldehyde dimethyl acetal, 98%, Acros Organics

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
40015-15-4
C5H12O2S
136.21
MFCD00009848
DVOAQLUDKIFSNB-UHFFFAOYSA-N
methylthio acetaldehyde dimethyl acetal,1,1-dimethoxy-2-methylthio ethane,ethane, 1,1-dimethoxy-2-methylthio,1,1-dimethoxy-2-methylsulfanyl-ethane,ethane,1,1-dimethoxy-2-methylthio,methylmercapto acetaldehyde dimethyl acetal,1,1-dimethoxy-2-methylsulfanyl ethane,acmc-209j9v,methylthio acetaldehyde dimethylacetal,2-methylthio acetaldehyde dimethyl acetal
638113
1,1-dimethoxy-2-methylsulfanylethane
COC(CSC)OC
40015-15-4
C5H12O2S
136.21
MFCD00009848
DVOAQLUDKIFSNB-UHFFFAOYSA-N
methylthio acetaldehyde dimethyl acetal,1,1-dimethoxy-2-methylthio ethane,ethane, 1,1-dimethoxy-2-methylthio,1,1-dimethoxy-2-methylsulfanyl-ethane,ethane,1,1-dimethoxy-2-methylthio,methylmercapto acetaldehyde dimethyl acetal,1,1-dimethoxy-2-methylsulfanyl ethane,acmc-209j9v,methylthio acetaldehyde dimethylacetal,2-methylthio acetaldehyde dimethyl acetal
638113
1,1-dimethoxy-2-methylsulfanylethane
COC(CSC)OC
Boiling Point 60°C (13.0mmHg)
Flash Point -18°C
Specific Gravity 1.021
Density 1.0210g/mL
Refractive Index 1.4510 to 1.4550 (20°C, 589nm)
Quantity 10g
Color Colorless
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 136.21
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material (Methylthio)acetaldehyde dimethyl acetal
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF

GHS Signal Word: Danger