Alfa Aesar™ Methylthiomethyl acetate, 95%

Manufacturer: Alfa Aesar™ H3231018

Quantity 50g

 View more versions of this product

Catalog No. AAH3231018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16437-69-7
    Molecular Formula C4H8O2S
    Linear Formula CH3CO2CH2SCH3
    Formula Weight 120.17
    MDL Number MFCD00075528
    UN Number UN3272
    Synonym Acetic acid methylthiomethyl ester
    Chemical Name or Material Methylthiomethyl acetate

    Specifications

    Formula Weight 120.17
    Molecular Formula C4H8O2S
    Linear Formula CH3CO2CH2SCH3
    MDL Number MFCD00075528
    UN Number UN3272
    Synonym Acetic acid methylthiomethyl ester
    Health Hazard 1 Warning
    Boiling Point 149°C
    Quantity 50g
    Flash Point 44°C (111°F)
    Odor Pungent
    Health Hazard 3 GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    CAS 16437-69-7
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material Methylthiomethyl acetate
    Assay 95%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit