Alfa Aesar™ Methylthiomethyl p-tolyl sulfone, 99%

Manufacturer: Alfa Aesar™ H3187706

Quantity 5g

 View more versions of this product

Catalog No. AAH3187706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 59662-65-6
    Molecular Formula C9H12O2S2
    Formula Weight 216.32
    MDL Number MFCD00010750
    Synonym 4-[(Methylthio)methylsulfonyl]toluene
    Chemical Name or Material Methylthiomethyl p-tolyl sulfone

    Specifications

    Formula Weight 216.32
    Molecular Formula C9H12O2S2
    MDL Number MFCD00010750
    CAS 59662-65-6
    Synonym 4-[(Methylthio)methylsulfonyl]toluene
    Melting Point 77° to 81°C
    Quantity 5g
    Chemical Name or Material Methylthiomethyl p-tolyl sulfone
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit