Learn More
Enzo Life SciencesSupplier Diversity Partner Methyltransferase activity kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI907025
The Methyltransferase activity kit is a homogeneous mix-and-read fluorescent assay for the determination of any S-adenosyl-L-methionine (AdoMet/SAM) dependent methyltransferase activity. It is suitable for end-point or kinetic read options, which is ideal for determining mechanism of action, kinetics, and screening candidate compounds. The assay is amendable to HTS and miniaturization. Applications: Fluorescent detection, HTS. Species Reactivity: Species independent. Applications: Activity assay. Contents: Black Microtiter plate, Reaction buffer, Assay buffer, Detection solution, Positive control
Catalog No. 50-658-608