Learn More
Enzo Life SciencesSupplier Diversity Partner Methyltransferase HT activity kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI907032
The Methyltransferase HT activity kit is a homogeneous mix-and-read fluorescent assay for the determination of any S-adenosyl-L-methionine (AdoMet/SAM) dependent methyltransferase activity. It is suitable for end-point or kinetic read options, which is ideal for determining mechanism of action, kinetics, and screening candidate compounds. This kit is ideal for high throughout (HT) applications as the SAM is provided separately giving you the flexibility to titrate the SAM to be in excess for your specific MT of interest. This is ideal for driving kinetics of low turnover enzymes and getting more utility by using less precious sample. Applications: Fluorescent detection, HTS. Species Reactivity: Species independent. Applications: Activity assay. Contents: Black Microtiter plate, SAM-Free Reaction buffer, Assay buffer, Detection solution, SAM, Positive control
Catalog No. 50-658-615