Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyltri-n-butylammonium bromide, 98%

Click to view available options
Quantity:
10g
50g
250g
37026-88-3
C13H30BrN
280.294
MFCD00043201
DHAWHVVWUNNONG-UHFFFAOYSA-M
tributylmethylammonium bromide,methyltri-n-butylammonium bromide,1-butanaminium, n,n-dibutyl-n-methyl-, bromide,methyltributylammonium bromide,methyl tributyl ammonium bromide,tributyl methyl azanium bromide,acmc-20ajfa,ksc493m4d,tributylmethylammonium bro-mide
10062191
tributyl(methyl)azanium;bromide
CCCC[N+](C)(CCCC)CCCC.[Br-]
37026-88-3
C13H30BrN
280.294
MFCD00043201
DHAWHVVWUNNONG-UHFFFAOYSA-M
tributylmethylammonium bromide,methyltri-n-butylammonium bromide,1-butanaminium, n,n-dibutyl-n-methyl-, bromide,methyltributylammonium bromide,methyl tributyl ammonium bromide,tributyl methyl azanium bromide,acmc-20ajfa,ksc493m4d,tributylmethylammonium bro-mide
10062191
tributyl(methyl)azanium;bromide
CCCC[N+](C)(CCCC)CCCC.[Br-]
Quantity 50g
Melting Point 120°C to 122°C
Beilstein 3704268
Formula Weight 280.29
Percent Purity 98%
Chemical Name or Material Methyltri-n-butylammonium bromide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning