Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltrichlorosilane, 98+%, ACROS Organics™

Click to view available options
Quantity:
10mL
500mL
2.5L
Packaging:
Glass bottle
75-79-6
CH3Cl3Si
149.48
JLUFWMXJHAVVNN-UHFFFAOYSA-N
methyltrichlorosilane,trichloro methyl silane,silane, trichloromethyl,methyl-trichlorsilan,methylsilicochloroform,trichloromethylsilicon,methylsilyl trichloride,monomethyltrichlorosilane,silane, methyltrichloro,trichlor-methylsilan
6399
trichloro(methyl)silane
C[Si](Cl)(Cl)Cl
75-79-6
CH3Cl3Si
149.48
JLUFWMXJHAVVNN-UHFFFAOYSA-N
methyltrichlorosilane,trichloro methyl silane,silane, trichloromethyl,methyl-trichlorsilan,methylsilicochloroform,trichloromethylsilicon,methylsilyl trichloride,monomethyltrichlorosilane,silane, methyltrichloro,trichlor-methylsilan
6399
trichloro(methyl)silane
C[Si](Cl)(Cl)Cl
Viscosity 0.48 mPa.s (20°C)
Melting Point -77.0°C
Boiling Point 66.0°C
Refractive Index 1.4100 to 1.4120
Specific Gravity 1.27
Quantity 2.5L
Color Colorless
Infrared Spectrum Authentic
Density 1.2700g/mL
Packaging Glass bottle
Flash Point −15°C
Assay Percent Range 98+%
Linear Formula CH3SiCl3
Fieser 11,553; 12,527
Solubility Information Solubility in water: reacts
Formula Weight 149.48
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material Methyltrichlorosilane
Show More Show Less