Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyltrimethoxysilane, 97+%

Intermediate used in coatings

Manufacturer:  Alfa Aesar™ 43070AE

Quantity 100mL

 View more versions of this product

Catalog No. AA43070AE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1185-55-3
136.22
UN2924
Methyltrimethoxysilane
Si(OCH3)3CH3
MFCD00008342
Trimethoxymethylsilane

Specifications

1185-55-3
Si(OCH3)3CH3
MFCD00008342
Trimethoxymethylsilane
1.3696
−78°C
8°C (46°F)
≥97%
Liquid
136.22
UN2924
0.955g/mL
102°C
100mL
Methyltrimethoxysilane

Safety and Handling

GHS H Statement
H225-H314-H318

GHS P Statement
P210-P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P310-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit