Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltrimethoxysilane, 97%, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass Bottle
1185-55-3
C4H12O3Si
136.22
BFXIKLCIZHOAAZ-UHFFFAOYSA-N
methyltrimethoxysilane,trimethoxy methyl silane,silane, trimethoxymethyl,union carbide a-163,silane, methyltrimethoxy,methyl trimethoxysilane,silane a-163,dynasylan mtms,unii-0hi0d71mci,methyl-trimethoxysilane
14456
trimethoxy(methyl)silane
CO[Si](C)(OC)OC
1185-55-3
C4H12O3Si
136.22
BFXIKLCIZHOAAZ-UHFFFAOYSA-N
methyltrimethoxysilane,trimethoxy methyl silane,silane, trimethoxymethyl,union carbide a-163,silane, methyltrimethoxy,methyl trimethoxysilane,silane a-163,dynasylan mtms,unii-0hi0d71mci,methyl-trimethoxysilane
14456
trimethoxy(methyl)silane
CO[Si](C)(OC)OC
Melting Point -50°C
Flash Point 11°C
Boiling Point 102°C
Specific Gravity 0.95
Viscosity 0.5 mPa.s (20°C)
Refractive Index 1.3695 to 1.3715
Packaging AcroSeal™ Glass Bottle
Density 0.9500g/mL
Quantity 1L
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3Si(OCH3)3
Solubility Information Solubility in water: decomposes
Formula Weight 136.22
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Methyltrimethoxysilane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-685-