Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltrioxorhenium(VII), 98%, Thermo Scientific Chemicals

Catalog No. AAL18478MC
Click to view available options
Quantity:
100 mg

Methyltrioxorhenium(VII) is used as a catalyst for various transformations such as olefin metathesis. It is associated with hydrogen peroxide and used as an oxidizing agent. It is used to prepare acid or ester from the corresponding terminalalkynes. It is a catalytic oxidant used with urea hydrogen peroxide to prepare nitrones. It reacts with 2-methylnaphthalene to get 2-methyl-1,4-naphthoquinone (Vitamin K3). Further, it catalyzes the reaction with aldehydes and diazoketones to get an alkene. In addition to this, it is used as an effective antioxidant under various conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltrioxorhenium(VII) is used as a catalyst for various transformations such as olefin metathesis. It is associated with hydrogen peroxide and used as an oxidizing agent. It is used to prepare acid or ester from the corresponding terminalalkynes. It is a catalytic oxidant used with urea hydrogen peroxide to prepare nitrones. It reacts with 2-methylnaphthalene to get 2-methyl-1,4-naphthoquinone (Vitamin K3). Further, it catalyzes the reaction with aldehydes and diazoketones to get an alkene. In addition to this, it is used as an effective antioxidant under various conditions.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70197-13-6
Molecular Formula CH3O3Re-
Molecular Weight (g/mol) 249.239
MDL Number MFCD00192332
InChI Key VZSXFJPZOCRDPW-UHFFFAOYSA-N
Synonym methyltrioxorhenium vii, methyltrioxorhenium, methylrhenium trioxide, unii-883d8rdd5q, methylrhenium vii trioxide, mto, methyltrioxorhenium mi, t-4-methyltrioxorhenium, methylrhenium trioxide vii
PubChem CID 2734010
IUPAC Name carbanide;trioxorhenium
SMILES [CH3-].O=[Re](=O)=O

Specifications

Melting Point 110°C to 115°C
Quantity 100 mg
Linear Formula CH3ReO3
UN Number UN1479
Beilstein 3927932
Solubility Information Soluble in water.
Formula Weight 249.24
Percent Purity 98%
Sensitivity Light and moisture sensitive
Chemical Name or Material Methyltrioxorhenium(VII)
Hazard Category H272
Hazard Statement GHS H Statement
H272
May intensify fire; oxidizer.
Precautionary Statement P210-P220-P221-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.