Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltriphenylphosphonium Bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
500g
2.5kg
Packaging:
Plastic bottle
1779-49-3
C19H18BrP
357.22
MFCD00011804
LSEFCHWGJNHZNT-UHFFFAOYSA-M
methyltriphenylphosphonium bromide,methyltriphenylphosphanium bromide,triphenylmethylphosphonium bromide,methyl triphenylphosphonium bromide,methyl triphenyl phosphonium bromide,phosphonium, methyltriphenyl-, bromide,methyltriphenylphosphoniumbromide,methyl-triphenyl-phosphonium bromide
74505
methyl(triphenyl)phosphanium;bromide
C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
1779-49-3
C19H18BrP
357.22
MFCD00011804
LSEFCHWGJNHZNT-UHFFFAOYSA-M
methyltriphenylphosphonium bromide,methyltriphenylphosphanium bromide,triphenylmethylphosphonium bromide,methyl triphenylphosphonium bromide,methyl triphenyl phosphonium bromide,phosphonium, methyltriphenyl-, bromide,methyltriphenylphosphoniumbromide,methyl-triphenyl-phosphonium bromide
74505
methyl(triphenyl)phosphanium;bromide
C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
for C-C Bond Formation (Olefination)
Packaging Plastic bottle
Flash Point >240°C
Infrared Spectrum Authentic
Melting Point 230.0°C to 234.0°C
Color White
Quantity 100g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula CH3P(C6H5)3Br
Fieser 01,697
Solubility Information Solubility in water: 400g/L (25°C). Other solubilities: 950g/L methanol (20°C),soluble in aliphatic alcohols and chloroform,practically insoluble in arom. hydrocarbons and,acetone
Formula Weight 357.22
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Methyltriphenylphosphonium bromide
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 217-218-9